(2R,3S,4S,5S)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate 97%

Supplier: Ambeed
Warning

Synonyms: β-L-Ribofuranose 1-Acetate 2,3,5-Tribenzoate

A110232-1G A110232-250MG A110232-25G A110232-5G
77189-018EA 19.57 USD
77189-018 77189-020 77189-022 77189-024
(2R,3S,4S,5S)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate 97%
(2R,3S,4S,5S)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate

Precaución: for Research Use Only

Formula: C₂₈H₂₄O₉
MW: 504.49 g/mol
MDL Number: MFCD00628242
CAS Number: 3080-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR