6-Bromoquinazolin-4-ol 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A109994-100G
A109994-10G
A109994-1G
A109994-250MG
A109994-5G
A109994-25G
77160-678EA
315.18
USD
77160-678
77160-680
77160-682
77160-684
77570-562
77683-007
6-Bromoquinazolin-4-ol 98%, Ambeed.Inc
6-Bromoquinazolin-4-ol
Precaución: For research use only.
Formula:
C₈H₅BrN₂O MW: 225.04 g/mol |
MDL Number:
MFCD00115131 CAS Number: 32084-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...