1-(Triisopropylsilyl)pyrrole 95%

Supplier: Ambeed
Warning

Synonyms: 1-(Triisopropylsilyl)-1H-pyrrole

A109882-100G A109882-10G A109882-1G A109882-25G A109882-5G A109882-250MG
77189-002EA 577.83 USD
77189-002 77189-004 77189-006 77189-008 77189-010 77745-816
1-(Triisopropylsilyl)pyrrole 95%
1-(Triisopropylsilyl)pyrrole

Precaución: for Research Use Only

Formula: C₁₃H₂₅NSi
MW: 223.44 g/mol
MDL Number: MFCD00054932
CAS Number: 87630-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR