Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoate 95%

Supplier: Ambeed
Warning

A109708-100MG A109708-1G A109708-250MG A109708-5G
77160-656EA 8 USD
77160-656 77160-658 77160-660 77891-796
Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoate 95%
Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₈BF₃O₄
MDL Number: MFCD19105387
CAS Number: 1045795-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR