3-Bromoquinoline-8-carboxylic acid 98%

Supplier: Ambeed
Warning

A109559-100MG A109559-250MG A109559-1G
77160-652EA 55 USD
77160-652 77160-654 77891-795
3-Bromoquinoline-8-carboxylic acid 98%
3-Bromoquinoline-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MDL Number: MFCD19382315
CAS Number: 1315366-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR