2-Bromo-4-methoxy-5-methylaniline 98%

Supplier: Ambeed
Warning

A109060-1G A109060-250MG
77160-618EA 83.43 USD
77160-618 77160-620
2-Bromo-4-methoxy-5-methylaniline 98%
2-Bromo-4-methoxy-5-methylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.08 g/mol
MDL Number: MFCD24639271
CAS Number: 328400-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR