1,2-Dibromo-4-methoxy-3,5,6-trimethylbenzene 98%

Supplier: Ambeed
Warning

A108860-10G A108860-1G A108860-250MG A108860-5G
77160-608EA 133 USD
77160-608 77160-610 77160-612 77160-614
1,2-Dibromo-4-methoxy-3,5,6-trimethylbenzene 98%
1,2-Dibromo-4-methoxy-3,5,6-trimethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂O
MDL Number: MFCD22370201
CAS Number: 1359986-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR