3-Bromo-1-benzofuran 97%

Supplier: Ambeed
Warning

A108737-25G A108737-5G A108737-1G
77188-948EA 157 USD
77188-948 77188-950 77570-502
3-Bromo-1-benzofuran 97%
3-Bromo-1-benzofuran

Precaución: for Research Use Only

Formula: C₈H₅BrO
MDL Number: MFCD01029425
CAS Number: 59214-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR