2-Isopropylthiazole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

A108624-100MG A108624-1G A108624-250MG A108624-5G
77188-940EA 168.92 USD
77188-940 77188-942 77188-944 77188-946
2-Isopropylthiazole-5-carbaldehyde 97%
2-Isopropylthiazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₉NOS
MW: 155.21 g/mol
MDL Number: MFCD14615182
CAS Number: 207675-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR