7-Fluoro-1-methyl-1H-indole-2,3-dione 97%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-1-methylindoline-2,3-dione

A108557-250MG A108557-25G A108557-5G
77160-602EA 13.39 USD
77160-602 77160-604 77160-606
7-Fluoro-1-methyl-1H-indole-2,3-dione 97%
7-Fluoro-1-methyl-1H-indole-2,3-dione

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.148 g/mol
MDL Number: MFCD21984607
CAS Number: 875003-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR