1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-(3-(Chloromethyl)-4-methoxyphenyl)ethanone, 3'-(Chloromethyl)-4'-methoxyacetophenone
A108330-1G
A108330-25G
A108330-5G
77188-930EA
12
USD
77188-930
77188-932
77188-934
1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone 98%
1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone
Precaución: for Research Use Only
Formula:
C₁₀H₁₁ClO₂ |
MDL Number:
MFCD00045267 CAS Number: 62581-82-2 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...