4-Amino-2-fluoro-5-methoxybenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A108277-100MG
A108277-250MG
77160-592EA
14
USD
77160-592
77160-594
4-Amino-2-fluoro-5-methoxybenzonitrile 97%
4-Amino-2-fluoro-5-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₇FN₂O MW: 166.152 g/mol |
MDL Number:
MFCD26407017 CAS Number: 1441723-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...