Dimethyl 4,5-dibromophthalate 97%

Supplier: Ambeed
Warning

A108270-100MG A108270-5G A108270-1G
77160-588EA 20.6 USD
77160-588 77160-590 77682-992
Dimethyl 4,5-dibromophthalate 97%
Dimethyl 4,5-dibromophthalate

Precaución: For research use only.

Formula: C₁₀H₈Br₂O₄
MW: 351.97 g/mol
MDL Number: MFCD11617187
CAS Number: 859299-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR