5,5'-Dimethyl-[1,1'-biphenyl]-2,2'-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A108109-100MG A108109-1G A108109-250MG
77160-578EA 22.66 USD
77160-578 77160-580 77160-582
5,5'-Dimethyl-[1,1'-biphenyl]-2,2'-dicarboxylic acid 95%
5,5'-Dimethyl-[1,1'-biphenyl]-2,2'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₄
MW: 270.28 g/mol
MDL Number: MFCD28123769
CAS Number: 93012-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR