2-(Piperidin-4-yl)-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Piperidinyl)benzimidazole

A107677-100G A107677-1G A107677-25G A107677-5G
77160-556EA 731 USD
77160-556 77160-558 77160-560 77160-562
2-(Piperidin-4-yl)-1H-benzo[d]imidazole 97%
2-(Piperidin-4-yl)-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃
MDL Number: MFCD01475843
CAS Number: 38385-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR