1-(5-Amino-2-pyridinyl)-4-piperidinol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(5-Aminopyridin-2-yl)piperidin-4-ol
A107604-100MG
A107604-1G
A107604-250MG
77188-892EA
57.68
USD
77188-892
77188-894
77188-896
1-(5-Amino-2-pyridinyl)-4-piperidinol 98%
1-(5-Amino-2-pyridinyl)-4-piperidinol
Precaución: for Research Use Only
Formula:
C₁₀H₁₅N₃O MW: 193.25 g/mol |
MDL Number:
MFCD09728697 CAS Number: 476342-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...