MLN2480 98%

Supplier: Ambeed
Warning

Synonyms: BIIB024, BIIB-024, (R)-2-(1-(6-amino-5-chloropyrimidine-4-carboxamido)ethyl)-N-(5-chloro-4-(trifluoromethyl)pyridin-2-yl)thiazole-5-carboxamide, 2-[(1R)-1-[(6-amino-5-chloropyrimidine-4-carbonyl)amino]ethyl]-N-[5-chloro-4-(trifluoromethyl)pyridin-2-yl]-1,3-thiazole-5-carboxamide, AMG2112819, AMG-2112819, Tovorafenib, TAK580, TAK 580, TAK-58, BIIB 024, ML-N2480, MLN2480, ML N2480, AMG 2112819

A107516-2MG A107516-5MG A107516-1MG
77160-550EA 29.87 USD
77160-550 77160-552 77682-985
MLN2480 98%
MLN2480

Precaución: For research use only.

Formula: C₁₇H₁₂Cl₂F₃N₇O₂S
MW: 506.29 g/mol
MDL Number: MFCD22571730
CAS Number: 1096708-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR