1,4-di-Boc-piperazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1,4-Di-Boc-piperazine-2-carboxylic acid

A107341-100G A107341-1G A107341-250MG A107341-25G A107341-5G
77188-856EA 440.84 USD
77188-856 77188-858 77188-860 77188-862 77188-864
1,4-di-Boc-piperazine-2-carboxylic acid 97%
1,4-di-Boc-piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₆N₂O₆
MW: 330.3800 g/mol
MDL Number: MFCD02179115
CAS Number: 181955-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR