4-Bromo-3-chloro-5-fluoroaniline 95%

Supplier: Ambeed
Warning

A107315-1G A107315-25G A107315-5G
77160-544EA 20 USD
77160-544 77160-546 77160-548
4-Bromo-3-chloro-5-fluoroaniline 95%
4-Bromo-3-chloro-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD28126799
CAS Number: 1297540-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR