5-Bromo-3-pyridineacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-pyridylacetic acid

A106935-100MG A106935-5G A106935-1G
77160-530EA 35.02 USD
77160-530 77160-532 77682-983
5-Bromo-3-pyridineacetic acid 98%
5-Bromo-3-pyridineacetic acid

Precaución: For research use only.

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD00829308
CAS Number: 39891-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR