(-)-Benzotetramisole ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (-)-BTM, (S)-2-Phenyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole, (2S)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole

A106904-100MG A106904-1G A106904-250MG A106904-5G
77160-522EA 24.72 USD
77160-522 77160-524 77160-526 77160-528
(-)-Benzotetramisole ≥98%, ee 99%
(-)-Benzotetramisole

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂S
MW: 252.34 g/mol
MDL Number: MFCD15072149
CAS Number: 950194-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR