4-(Methylsulfonyl)phenol 98%

Supplier: Ambeed
Warning

A106800-100G A106800-25G A106800-5G
77188-838EA 126.69 USD
77188-838 77188-840 77188-842
4-(Methylsulfonyl)phenol 98%
4-(Methylsulphonyl)phenol

Precaución: for Research Use Only

Formula: C₇H₈O₃S
MW: 172.20 g/mol
MDL Number: MFCD00047806
CAS Number: 14763-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR