1-Bromo-2-fluoro-3-iodobenzene 97%

Supplier: Ambeed
Warning

A106737-100MG A106737-1G A106737-250MG A106737-25G A106737-5G
77188-828EA 16.48 USD
77188-828 77188-830 77188-832 77188-834 77188-836
1-Bromo-2-fluoro-3-iodobenzene 97%
1-Bromo-2-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MW: 300.895 g/mol
MDL Number: MFCD13194483
CAS Number: 958458-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR