1,4-Dioxaspiro[4.5]decane-8-carbonitrile 97%

Supplier: Ambeed
Danger

A106670-10G A106670-1G A106670-250MG A106670-5G A106670-25G
77160-496EA 113 USD
77160-496 77160-498 77160-500 77160-502 77891-783
1,4-Dioxaspiro[4.5]decane-8-carbonitrile 97%
1,4-Dioxaspiro[4.5]decane-8-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MDL Number: MFCD09037879
CAS Number: 69947-09-7
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR