Disodium naphthalene-1,5-disulphonate 98%

Supplier: Ambeed
Warning

Synonyms: Disodium 1,5-Naphthalenedisulphonate hydrate, Naphthalene-1,5-disulphonic acid disodium salt, 1,5-Naphthalenedisulphonic acid disodium salt, Disodium 1,5-naphthalenedisulphonate, Sodium 1,5-naphthalenedisulphonate dibasic

A106566-1000G A106566-100G A106566-25G A106566-500G
77188-816EA 43 USD
77188-816 77188-818 77188-820 77188-822
Disodium naphthalene-1,5-disulphonate 98%
Disodium naphthalene-1,5-disulphonate

Precaución: for Research Use Only

Formula: C₁₀H₆Na₂O₆S₂
MDL Number: MFCD00064178
CAS Number: 1655-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR