(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A106473-100MG
A106473-1G
A106473-250MG
77160-488EA
9.27
USD
77160-488
77160-490
77160-492
(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride 97%
(2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₀ClNO₃ MW: 167.59 g/mol |
MDL Number:
MFCD09908038 CAS Number: 32968-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...