(S)-tert-Butyl(2-oxopiperidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl (2-oxopiperidin-3-yl)carbamate, (3S)-3-Aminopiperidin-2-one, 3-BOC protected

A106349-10G A106349-250MG A106349-5G A106349-25G
77188-812EA 189.52 USD
77188-812 77188-814 77570-376 77745-802
(S)-tert-Butyl(2-oxopiperidin-3-yl)carbamate 95%
(S)-tert-Butyl(2-oxopiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.2600 g/mol
MDL Number: MFCD08166304
CAS Number: 92235-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR