1-(5-Chloro-2-methylphenyl)propan-1-ol 97%

Supplier: Ambeed
Warning

A106205-100MG A106205-250MG
77188-808EA 49.44 USD
77188-808 77188-810
1-(5-Chloro-2-methylphenyl)propan-1-ol 97%
1-(5-Chloro-2-methylphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClO
MW: 184.66 g/mol
MDL Number: MFCD11521240
CAS Number: 1314976-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR