1,5-Dibromo-2,4-dimethoxybenzene 97%

Supplier: Ambeed
Warning

A106185-1G A106185-5G A106185-25G
77188-804EA 10.3 USD
77188-804 77188-806 77565-856
1,5-Dibromo-2,4-dimethoxybenzene 97%
1,5-Dibromo-2,4-dimethoxybenzene

Precaución: For research use only.

Formula: C₈H₈Br₂O₂
MW: 295.956 g/mol
MDL Number: MFCD00061097
CAS Number: 24988-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR