Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-β-HomoAsp(OtBu)-OH, Fmoc-β-HoAsp(OtBu)-OH
A106179-1G
A106179-5G
A106179-250MG
A106179-25G
77160-480EA
91.67
USD
77160-480
77549-038
77682-977
77682-978
Fmoc-L-β-homoaspartic acid-gamma-t-butyl ester 98%
Fmoc-L-beta-homoaspartic acid-gamma-t-butyl ester
Precaución: For research use only.
Formula:
C₂₄H₂₇NO₆ MW: 425.49 g/mol |
MDL Number:
MFCD01862860 CAS Number: 209252-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...