tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-aminopiperidine-1-carboxylate hydrochloride, 4-Amino-1-Boc-piperidine hydrochloride, tert-Butyl 4-amino-1-piperidinecarboxylate hydrochloride

A105977-1G A105977-5G
77188-784EA 13 USD
77188-784 77188-786
tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride 95%
tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O₂
MW: 236.7400 g/mol
MDL Number: MFCD01321014
CAS Number: 189819-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR