1,4-Diiodobutane 97% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
Tetramethylene diiodide
Stabilization:
Stabilised with copper chip
A105370-100G
A105370-10G
A105370-1G
A105370-25G
A105370-500G
A105370-5G
77188-742EA
60
USD
77188-742
77188-744
77188-746
77188-748
77188-750
77188-752
1,4-Diiodobutane 97% stabilised
1,4-Diiodobutane
Precaución: for Research Use Only
Formula:
C₄H₈I₂ MW: 309.92 g/mol Boiling Pt: 247 Melting Pt: 5.8 |
MDL Number:
MFCD00001099 CAS Number: 628-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...