1,1-Bis(4-methoxyphenyl)prop-2-yn-1-ol 97%

Supplier: Ambeed
Warning

A105287-1G A105287-250MG A105287-5G A105287-25G
77188-736EA 21.63 USD
77188-736 77188-738 77188-740 77745-797
1,1-Bis(4-methoxyphenyl)prop-2-yn-1-ol 97%
1,1-Bis(4-methoxyphenyl)prop-2-yn-1-ol

Precaución: for Research Use Only

Formula: C₁₇H₁₆O₃
MW: 268.31 g/mol
MDL Number: MFCD00420323
CAS Number: 101597-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR