5-Bromo-2-chloro-4-methylpyridine 97%

Supplier: Ambeed
Warning

A105216-100G A105216-10G A105216-1G A105216-25G A105216-500G A105216-5G
77160-448EA 119.48 USD
77160-448 77160-450 77160-452 77160-454 77160-456 77160-458
5-Bromo-2-chloro-4-methylpyridine 97%
5-Bromo-2-chloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MW: 206.47 g/mol
MDL Number: MFCD03427663
CAS Number: 778611-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR