2-Amino-5-ethyl-6-hydroxypyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A105165-1G A105165-250MG
77160-444EA 88.58 USD
77160-444 77160-446
2-Amino-5-ethyl-6-hydroxypyrimidin-4(3H)-one 97%
2-Amino-5-ethyl-6-hydroxypyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂
MW: 155.15 g/mol
MDL Number: MFCD00090758
CAS Number: 30201-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR