Sodium pyrene-1,3,6,8-tetrasulfonate 98% (contains < 15% H₂O)

Supplier: Ambeed
Warning

A105036-100G A105036-25G A105036-500G A105036-5G
77188-718EA 113.3 USD
77188-718 77188-720 77188-722 77188-724
Sodium pyrene-1,3,6,8-tetrasulfonate 98% (contains < 15% H₂O)
Sodium pyrene-1,3,6,8-tetrasulfonate

Precaución: for Research Use Only

Formula: C₁₆H₆Na₄O₁₂S₄
MW: 610.43 g/mol
MDL Number: MFCD00042029
CAS Number: 59572-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR