2-Amino-3-chloro-5-fluorobenzoic acid 95%

Supplier: Ambeed
Warning

A104785-100MG A104785-1G A104785-250MG
77160-426EA 8.24 USD
77160-426 77160-428 77160-430
2-Amino-3-chloro-5-fluorobenzoic acid 95%
2-Amino-3-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClFNO₂
MW: 189.57 g/mol
MDL Number: MFCD11193640
CAS Number: 1022961-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR