1-(3-Amino-4-chlorophenyl)ethanone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(3-Amino-4-chlorophenyl)ethan-1-one, 3'-Amino-4'-chloroacetophenone
A104780-10G
A104780-1G
A104780-250MG
77188-706EA
100.94
USD
77188-706
77188-708
77188-710
1-(3-Amino-4-chlorophenyl)ethanone 97%
1-(3-Amino-4-chlorophenyl)ethanone
Precaución: for Research Use Only
Formula:
C₈H₈ClNO MW: 169.61 g/mol |
MDL Number:
MFCD00122619 CAS Number: 79406-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...