2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-allylpent-4-enoic acid 95%

Supplier: Ambeed
Warning

A104665-1G A104665-250MG A104665-5G
77160-404EA 182.31 USD
77160-404 77160-406 77160-408
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-allylpent-4-enoic acid 95%
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-allylpent-4-enoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₃NO₄
MW: 377.43 g/mol
MDL Number: MFCD17215634
CAS Number: 1311992-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR