4-Chloro-6-iodoquinoline-3-carbonitrile 95%

Supplier: Ambeed
Warning

A104559-100MG A104559-1G A104559-250MG
77160-398EA 44.29 USD
77160-398 77160-400 77160-402
4-Chloro-6-iodoquinoline-3-carbonitrile 95%
4-Chloro-6-iodoquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₄ClIN₂
MW: 314.50 g/mol
MDL Number: MFCD11194094
CAS Number: 214483-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR