Butyl isobutyl phthalate 95%

Supplier: Ambeed
Danger

A104493-1G A104493-250MG
77160-394EA 349.17 USD
77160-394 77160-396
Butyl isobutyl phthalate 95%
Butyl isobutyl phthalate

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₄
MW: 278.34 g/mol
MDL Number: MFCD11114531
CAS Number: 17851-53-5
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR