1,1-Bis(diphenylphosphino)ethylene 98%

Supplier: Ambeed
Warning

Synonyms: Ethene-1,1-diylbis(diphenylphosphine)

A104462-1G A104462-250MG A104462-5G
77188-700EA 124.63 USD
77188-700 77188-702 77188-704
1,1-Bis(diphenylphosphino)ethylene 98%
1,1-Bis(diphenylphosphino)ethylene

Precaución: for Research Use Only

Formula: C₂₆H₂₂P₂
MW: 396.41 g/mol
MDL Number: MFCD00191809
CAS Number: 84494-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR