Methyl 4-amino-3-isopropoxybenzoate 97%

Supplier: Ambeed
Warning

A104134-1G A104134-250MG
77188-692EA 216.3 USD
77188-692 77188-694
Methyl 4-amino-3-isopropoxybenzoate 97%
Methyl 4-amino-3-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.24 g/mol
MDL Number: MFCD12168764
CAS Number: 681465-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR