(R)-tert-Butyl 3-amino-4-phenylbutanoate hydrochloride 95%

Supplier: Ambeed
Warning

A104066-100MG A104066-1G A104066-250MG
77188-686EA 39.14 USD
77188-686 77188-688 77188-690
(R)-tert-Butyl 3-amino-4-phenylbutanoate hydrochloride 95%
(R)-tert-Butyl 3-amino-4-phenylbutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₂ClNO₂
MW: 271.78 g/mol
MDL Number: MFCD29038839
CAS Number: 422324-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR