6-Chloro-4-methoxypyridine-3-carbaldehyde 97%

Supplier: Ambeed
Warning

A103964-100MG A103964-1G A103964-250MG
77188-680EA 40 USD
77188-680 77188-682 77188-684
6-Chloro-4-methoxypyridine-3-carbaldehyde 97%
6-Chloro-4-methoxypyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MDL Number: MFCD18255526
CAS Number: 1256823-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR