TFA-ap-dC 97%

Supplier: Ambeed
Warning

A103724-100MG A103724-1G
77160-354EA 44.29 USD
77160-354 77160-356
TFA-ap-dC 97%
TFA-ap-dC

Precaución: for Research Use Only

Formula: C₁₄H₁₅F₃N₄O₅
MW: 376.29 g/mol
MDL Number: MFCD28134647
CAS Number: 115899-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR