2-(2-Methylphenyl)propanenitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-(o-Tolyl)propanenitrile

A103714-1G A103714-250MG
77160-350EA 43.26 USD
77160-350 77160-352
2-(2-Methylphenyl)propanenitrile 95%
2-(2-Methylphenyl)propanenitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₁N
MW: 145.201 g/mol
MDL Number: MFCD11036533
CAS Number: 58422-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR