Ethyl 2-phenylthiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A103712-1G A103712-250MG
77160-344EA 41.2 USD
77160-344 77160-346 77160-348
Ethyl 2-phenylthiazole-5-carboxylate 97%
Ethyl 2-phenylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂S
MDL Number: MFCD16036490
CAS Number: 172678-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR