1,2-Dibromotetrafluorobenzene 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dibromo-3,4,5,6-tetrafluorobenzene

A103640-100G A103640-10G A103640-1G A103640-250MG A103640-25G A103640-5G
77188-664EA 574.99 USD
77188-664 77188-666 77188-668 77188-670 77188-672 77188-674
1,2-Dibromotetrafluorobenzene 95%
1,2-Dibromotetrafluorobenzene

Precaución: for Research Use Only

Formula: C₆Br₂F₄
MW: 307.87 g/mol
Melting Pt: 12
MDL Number: MFCD00039209
CAS Number: 827-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR