4-Chloro-3-(trifluoromethyl)benzaldehyde 95%

Supplier: Ambeed
Warning

A103482-10G A103482-1G A103482-25G A103482-5G
77188-656EA 25.75 USD
77188-656 77188-658 77188-660 77188-662
4-Chloro-3-(trifluoromethyl)benzaldehyde 95%
4-Chloro-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
MW: 208.57 g/mol
MDL Number: MFCD00040722
CAS Number: 34328-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR